siRNA Details
Gene Symbol: Ube2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145420.2 | NP_663395.1 | 6 | 509 | ||
GenBank mRNA | AK144468.1 | - | 6 | 464 | ||
AK148364.1 | - | 6 | 497 | |||
BC019464.1 | - | 6 | 480 | 4205941 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
