siRNA Details
Gene Symbol: Tmem259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003949.3 | NP_001003949.2 | 11 | 1509 | ||
GenBank mRNA | AK131170.1 | - | 226 | |||
AY096036.1 | - | 11 | 1219 | |||
AY096037.1 | - | 10 | 1291 | |||
BC005494.1 | - | 10 | 1032 | |||
BC052787.1 | - | 10 | 1051 | |||
BC056654.1 | - | 2 | 362 | |||
BC080302.1 | - | 712 | ||||
BC100297.1 | - | 8 | 646 |

More Information
Other Species Targeted

Gene Ontology Categories:
