siRNA Details
Gene Symbol: Tmem259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003949.3 | NP_001003949.2 | 7 | 1133 | ||
GenBank mRNA | AY096036.1 | - | 7 | 843 | ||
AY096037.1 | - | 7 | 843 | |||
BC005494.1 | - | 6 | 656 | |||
BC052787.1 | - | 6 | 675 | |||
BC080302.1 | - | 193 | ||||
BC100297.1 | - | 4 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
