siRNA Details
Gene Symbol: Abhd17a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145421.2 | NP_663396.1 | 4, 5 | 1066 | ||
GenBank mRNA | AK220316.1 | - | 2, 3 | 4313 | ||
BC005632.1 | - | 4, 5 | 1041 | 3964094 | ||
BC082997.1 | - | 4, 5 | 1030 | 4160194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
