siRNA Details
Gene Symbol: Taf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020024.3 | NP_064408.2 | 3, 4 | 476 | ||
GenBank mRNA | AK047115.1 | - | 478 | |||
AK139383.1 | - | 486 | ||||
BC031418.1 | - | 3, 4 | 476 | 3667894 | ||
BC080311.1 | - | 3, 4 | 477 | 5713860 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001134735, BC168203.
Gene Ontology Categories:
