siRNA Details
Gene Symbol: Taf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020024.3 | NP_064408.2 | 4, 5 | 596 | ||
GenBank mRNA | AK047115.1 | - | 598 | |||
AK139383.1 | - | 606 | ||||
BC031418.1 | - | 4, 5 | 596 | 3667894 | ||
BC080311.1 | - | 4, 5 | 597 | 5713860 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
