siRNA Details
Gene Symbol: Ckap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175451.1 | NP_780660.1 | 2 | 563 | ||
GenBank mRNA | AK030708.1 | - | 2 | 447 | ||
AK132136.1 | - | 737 | ||||
BC025522.1 | - | 2 | 214 | |||
BC138318.1 | - | 3 | 585 | 8861338 | ||
BC138320.1 | - | 3 | 585 | 8861340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
