siRNA Details
Gene Symbol: Slc5a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145423.2 | NP_663398.2 | 14 | 2000 | ||
GenBank mRNA | AK039927.1 | - | 14 | 1829 | ||
AK132487.1 | - | 2 | 160 | |||
AK136457.1 | - | 14 | 2010 | |||
AY484428.1 | - | 14 | 2000 | |||
BC017691.1 | - | 1955 | 4239161 |

More Information
Other Species Targeted

Gene Ontology Categories:
