siRNA Details
Gene Symbol: Os9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171026.1 | NP_001164497.1 | 14 | 1895 | ||
NM_177614.3 | NP_808282.2 | 13 | 1730 | |||
GenBank mRNA | AK028047.1 | - | 14 | 1872 | ||
AK185854.1 | - | 2 | 218 | |||
BC006844.1 | - | 13 | 1550 | |||
BC031768.1 | - | 13 | 1695 | 4487829 | ||
DV647242.1 | - | 3 | 574 | |||
DV660362.1 | - | 2 | 186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
