siRNA Details
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290634.1 | NP_001277563.1 | 8, 9 | 1553 | ||
NM_139297.6 | NP_647458.1 | 8, 9 | 1743 | |||
GenBank mRNA | AF424698.1 | - | 8, 9 | 1591 | ||
AK033445.1 | - | 8, 9 | 1590 | |||
AK153885.1 | - | 7, 8 | 2686 | |||
AK205646.1 | - | 1, 2 | 225 | |||
AK206548.1 | - | 1, 2 | 214 | |||
AK211484.1 | - | 1, 2 | 221 | |||
AK215699.1 | - | 1, 2 | 203 | |||
AK216282.1 | - | 1, 2 | 193 | |||
AK217204.1 | - | 1, 2 | 224 | |||
BC023779.1 | - | 1, 2 | 192 | |||
BC023810.1 | - | 8, 9 | 1594 | 5353993 | ||
BC025585.1 | - | 8, 9 | 1498 | 5324928 | ||
BC026626.1 | - | 7, 8 | 1280 | |||
BC061208.1 | - | 8, 9 | 1630 | 30250244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
