siRNA Details
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290634.1 | NP_001277563.1 | 5, 6 | 817 | ||
NM_139297.6 | NP_647458.1 | 5, 6 | 1007 | |||
GenBank mRNA | AF424698.1 | - | 5, 6 | 855 | ||
AK033445.1 | - | 5, 6 | 854 | |||
AK153885.1 | - | 4, 5 | 1950 | |||
AK218540.1 | - | 2, 3 | 175 | |||
BC023810.1 | - | 5, 6 | 858 | 5353993 | ||
BC025585.1 | - | 5, 6 | 762 | 5324928 | ||
BC026626.1 | - | 4, 5 | 544 | |||
BC061208.1 | - | 5, 6 | 894 | 30250244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
