siRNA Details
Gene Symbol: Ehbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252515.1 | NP_001239444.1 | 14 | 1974 | ||
NM_153078.4 | NP_694718.4 | 14 | 1899 | |||
GenBank mRNA | AF424697.1 | - | 13 | 1749 | ||
AK173055.1 | - | 14 | 1975 |
