siRNA Details
Gene Symbol: Dhrs7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172112.1 | NP_001165583.1 | 6 | 829 | ||
NM_145428.2 | NP_663403.1 | 6 | 800 | |||
GenBank mRNA | AK052209.1 | - | 6 | 752 | ||
AK139084.1 | - | 6 | 781 | |||
AK144057.1 | - | 6 | 735 | |||
BC003479.1 | - | 6 | 755 | 2655855 | ||
JX196872.1 | - | 6 | 719 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
