siRNA Details
Gene Symbol: Kdm6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017426.1 | NP_001017426.1 | 18 | 4645 | ||
GenBank mRNA | AK129121.1 | - | 8 | 1437 | ||
AK136019.1 | - | 8 | 826 | |||
BC038313.1 | - | 3 | 283 | |||
BC075632.1 | - | 19 | 4821 | 30362899 |

More Information
Other Species Targeted

Gene Ontology Categories:
