siRNA Details
Gene Symbol: Camta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190376.1 | NP_001177305.1 | 21 | 3628 | ||
NM_001190378.1 | NP_001177307.1 | 20 | 3607 | |||
NM_001190379.1 | NP_001177308.1 | 19 | 3535 | |||
NM_178116.4 | NP_835217.1 | 21 | 3643 | |||
GenBank mRNA | AK122400.1 | - | 19 | 3284 | ||
AK150186.1 | - | 20 | 3461 | |||
AK169118.1 | - | 19 | 3374 | |||
BC023962.1 | - | 15 | 3421 | |||
BC027039.1 | - | 10 | 1352 | |||
BC038079.1 | - | 3 | 234 | |||
BC049133.1 | - | 21 | 3472 | 6827800 | ||
BC056395.1 | - | 21 | 3457 | 6852349 | ||
BC059065.1 | - | 19 | 3402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
