siRNA Details
Gene Symbol: Gm525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033266.2 | NP_001028438.1 | 3 | 307 | ||
GenBank mRNA | AA237157.1 | - | 3 | 203 | ||
AK040024.1 | - | 3 | 225 | |||
AW823710.1 | - | 3 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
