siRNA Details
Gene Symbol: Scrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146027.2 | NP_666139.1 | 2 | 186 | ||
GenBank mRNA | AK075869.1 | - | 2 | 150 | ||
BC010784.1 | - | 2 | 137 | |||
BC021346.1 | - | 2 | 155 | 4163541 | ||
CN691874.1 | - | 2 | 186 |

More Information
Other Species Targeted

Gene Ontology Categories:
