siRNA Details
Gene Symbol: Gpr179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081220.1 | NP_001074689.1 | 1 | 666 | ||
GenBank mRNA | AK030625.1 | - | 1 | 670 | ||
AK038894.1 | - | 1 | 660 |

More Information
Other Species Targeted

Gene Ontology Categories:
