siRNA Details
Gene Symbol: Stac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146028.4 | NP_666140.1 | 2 | 610 | ||
GenBank mRNA | AJ608761.1 | - | 2 | 423 | ||
AK078132.1 | - | 2 | 611 | |||
AK079357.1 | - | 2 | 468 | |||
AK144433.1 | - | 2 | 466 | |||
BC024864.1 | - | 2 | 516 | 5361431 | ||
BC029794.1 | - | 2 | 457 | 4949472 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
