siRNA Details
Gene Symbol: Nr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145434.4 | NP_663409.2 | 7 | 2087 | ||
GenBank mRNA | AK041047.1 | - | 7 | 1943 | ||
AK135640.1 | - | 7 | 1949 | |||
AK137398.1 | - | 7 | 1974 | |||
AK137582.1 | - | 7 | 1952 | |||
AK146430.1 | - | 7 | 2044 | |||
AK154931.1 | - | 7 | 2088 | |||
AK155597.1 | - | 7 | 2088 | |||
BC008989.1 | - | 7 | 1532 | 3585478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
