siRNA Details
Gene Symbol: Nags | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145829.2 | NP_665828.1 | 3 | 927 | ||
GenBank mRNA | AF462069.1 | - | 3 | 883 | ||
AJ489814.1 | - | 3 | 853 | |||
AK075765.1 | - | 2 | 573 | |||
BC036161.1 | - | 2 | 1025 | |||
BC057990.1 | - | 3 | 891 | 5100021 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001107053, XM_008768055, XM_006247328.
Gene Ontology Categories:
