siRNA Details
Gene Symbol: Nags | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145829.2 | NP_665828.1 | 7 | 1593 | ||
GenBank mRNA | AF462069.1 | - | 7 | 1549 | ||
AJ489814.1 | - | 7 | 1519 | |||
AK075765.1 | - | 6 | 1239 | |||
BC036161.1 | - | 4 | 2244 | |||
BC057990.1 | - | 7 | 1557 | 5100021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
