siRNA Details
Gene Symbol: Atxn7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098836.1 | NP_001092306.1 | 7 | 809 | ||
NM_001098837.1 | NP_001092307.1 | 7 | 788 | |||
GenBank mRNA | AK090368.1 | - | 7 | 790 | ||
BC059880.1 | - | 1 | 147 | |||
BC130269.1 | - | 6 | 559 | |||
BM950535.1 | - | 5 | 366 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
