siRNA Details
Gene Symbol: Cdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285988.1 | NP_001272917.1 | 14 | 2116 | ||
NM_001285989.1 | NP_001272918.1 | 14 | 2098 | |||
NM_001285990.1 | NP_001272919.1 | 13 | 2021 | |||
NM_145436.2 | NP_663411.2 | 14 | 2101 | |||
GenBank mRNA | AK148278.1 | - | 14 | 1956 | ||
AK161143.1 | - | 3 | 477 | |||
AK161471.1 | - | 14 | 1967 | |||
BC157955.1 | - | 14 | 2115 | 9088128 | ||
BC172100.1 | - | 14 | 2115 | 9054386 | ||
BC172149.1 | - | 13 | 2020 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
