siRNA Details
Gene Symbol: Cd300a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170758.3 | NP_739564.1 | 2 | 520 | ||
GenBank mRNA | AB065156.1 | - | 2 | 239 | ||
AB091765.1 | - | 2 | 258 | |||
AB091766.1 | - | 2 | 242 | |||
AK045869.1 | - | 2 | 520 | |||
AK053160.1 | - | 2 | 208 | |||
AY457054.1 | - | 2 | 196 | |||
BC139296.1 | - | 2 | 456 | 8862318 | ||
BC139297.1 | - | 2 | 456 | 8862319 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
