siRNA Details
Gene Symbol: Cd300ld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145437.2 | NP_663412.1 | 2 | 261 | ||
GenBank mRNA | AK028662.1 | - | 2 | 273 | ||
AK041292.1 | - | 2 | 297 | |||
AY457051.1 | - | 2 | 185 | |||
BC019814.1 | - | 2 | 261 | 5125383 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
