siRNA Details
Gene Symbol: Unk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286006.1 | NP_001272935.1 | 3 | 437 | ||
NM_172569.4 | NP_766157.1 | 3 | 437 | |||
GenBank mRNA | AK046395.1 | - | 3 | 432 | ||
AK077635.1 | - | 3 | 438 | |||
AK078095.1 | - | 3 | 428 | |||
AK129438.1 | - | 3 | 415 | |||
AK134361.1 | - | 3 | 557 | |||
BB619163.1 | - | 3 | 438 | |||
BC039964.1 | - | 3 | 399 | |||
BC054452.1 | - | 3 | 374 | 6491704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
