siRNA Details
Gene Symbol: Rnf157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027258.1 | NP_081534.1 | 3 | 513 | ||
GenBank mRNA | BC053070.1 | - | 3 | 292 | ||
CF539215.1 | - | 2 | 155 |

More Information
Other Species Targeted

Gene Ontology Categories:
