siRNA Details
Gene Symbol: Tnrc6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198022.2 | NP_932139.2 | 17 | 5107 | ||
GenBank mRNA | AK036989.1 | - | 13 | 2586 | ||
AK122529.1 | - | 13 | 3058 | |||
AK147471.1 | - | 18 | 5105 | |||
AK147551.1 | - | 16 | 7982 |

More Information
Other Species Targeted

Gene Ontology Categories:
