siRNA Details
Gene Symbol: Lrrc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153545.2 | NP_705773.2 | 7 | 860 | ||
GenBank mRNA | AK081566.1 | - | 7 | 859 | ||
AK153361.1 | - | 7 | 861 | |||
BC023296.1 | - | 7 | 840 | 3662961 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
