siRNA Details
Gene Symbol: Pqlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172574.2 | NP_766162.2 | 5, 6 | 631 | ||
GenBank mRNA | AK053171.1 | - | 5, 6 | 580 | ||
AK150094.1 | - | 5, 6 | 543 | |||
BC025220.1 | - | 5, 6 | 569 | 5025905 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
