siRNA Details
Gene Symbol: Gphb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175644.3 | NP_783575.2 | 3 | 343 | ||
GenBank mRNA | AF467771.1 | - | 2 | 229 | ||
AK078727.1 | - | 3 | 345 | |||
BC111115.1 | - | 2 | 184 | |||
BC139462.1 | - | 3 | 283 | 8862484 | ||
BC139463.1 | - | 3 | 283 | 8862485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
