siRNA Details
Gene Symbol: Fam161b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172581.2 | NP_766169.1 | 6, 7 | 1644 | ||
GenBank mRNA | AK034254.1 | - | 6, 7 | 1647 | ||
AK036740.1 | - | 6, 7 | 1645 | |||
AK046195.1 | - | 5, 6 | 2666 | |||
BC062132.1 | - | 6, 7 | 1637 | 6509090 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
