siRNA Details
Gene Symbol: Nrde2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290303.1 | NP_001277232.1 | 11 | 2808 | ||
NM_183155.3 | NP_898978.2 | 11 | 2793 | |||
GenBank mRNA | BC002230.1 | - | 1 | 502 | ||
BC025577.1 | - | 6 | 1413 | 5323598 | ||
BC051175.1 | - | 10 | 2698 | |||
CN710548.1 | - | 1 | 323 |
