siRNA Details
Gene Symbol: Rcor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198023.2 | NP_932140.1 | 7 | 599 | ||
GenBank mRNA | AB093210.1 | - | 3 | 300 | ||
AK077445.1 | - | 1 | 40 | |||
BC042731.1 | - | 7 | 599 | 4936863 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
