siRNA Details
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081170.1 | NP_001074639.1 | 18 | 1958 | ||
NM_001291444.1 | NP_001278373.1 | 18 | 1964 | |||
NM_001291445.1 | NP_001278374.1 | 17 | 1868 | |||
GenBank mRNA | AK030003.1 | - | 13 | 1327 | ||
AK036581.1 | - | 17 | 1844 | |||
AK054000.1 | - | 18 | 1939 | |||
AK122326.1 | - | 16 | 1647 | |||
BC059076.1 | - | 3 | 528 |

More Information
Other Species Targeted

Gene Ontology Categories:
