siRNA Details
Gene Symbol: Cdca7l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146040.1 | NP_666152.1 | 3 | 277 | ||
GenBank mRNA | AK149673.1 | - | 3 | 255 | ||
BC006933.1 | - | 3 | 277 | 3602150 | ||
BC071198.1 | - | 3 | 244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
