siRNA Details
Gene Symbol: Larp4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172585.2 | NP_766173.1 | 7 | 848 | ||
GenBank mRNA | AK042491.1 | - | 7 | 846 | ||
AK080953.1 | - | 7 | 872 | |||
AK160759.1 | - | 7 | 849 | |||
AK172916.1 | - | 7 | 830 | |||
BC059218.1 | - | 7 | 828 | |||
BC078640.1 | - | 7 | 771 | 6506267 | ||
BC086626.1 | - | 7 | 828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
