siRNA Details
Gene Symbol: Pou6f2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175006.2 | NP_778171.2 | 1 | 364 | ||
GenBank mRNA | AK083772.1 | - | 1 | 364 | ||
BC094915.1 | - | 1 | 379 | 6435959 | ||
BC138151.1 | - | 1 | 283 | 8861170 | ||
BG807013.1 | - | 1 | 14 |
