siRNA Details
Gene Symbol: Amph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289546.1 | NP_001276475.1 | 6 | 555 | ||
NM_175007.2 | NP_778172.1 | 6 | 555 | |||
GenBank mRNA | AK047144.1 | - | 6 | 509 | ||
AK132168.1 | - | 6 | 556 | |||
BC054718.1 | - | 6 | 535 | 5686478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
