siRNA Details
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301781.1 | NP_001288710.1 | 8 | 2011 | ||
NM_207232.2 | NP_997115.1 | 7 | 1828 | |||
GenBank mRNA | AK030415.1 | - | 7 | 1821 | ||
AK038922.1 | - | 7 | 1741 | |||
AK053480.1 | - | 7 | 1599 | |||
AK132781.1 | - | 8 | 1920 | |||
BC066081.1 | - | 7 | 1729 | 30355705 |
