siRNA Details
Gene Symbol: Ice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144837.3 | NP_659086.2 | 17, 18 | 6450 | ||
GenBank mRNA | AK129244.1 | - | 5, 6 | 4887 | ||
AK134564.1 | - | 5, 6 | 2893 | |||
BC018507.1 | - | 4, 5 | 545 | |||
BC026613.1 | - | 5, 6 | 976 |

More Information
Other Species Targeted

Gene Ontology Categories:
