siRNA Details
Gene Symbol: Ice1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144837.3 | NP_659086.2 | 13 | 4216 | ||
GenBank mRNA | AK129244.1 | - | 1 | 2653 | ||
AK134564.1 | - | 1 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
