siRNA Details
Gene Symbol: Btf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170540.1 | NP_001164011.1 | 4 | 491 | ||
NM_145455.3 | NP_663430.2 | 4 | 693 | |||
GenBank mRNA | AK139664.1 | - | 4 | 466 | ||
AK146332.1 | - | 4 | 479 | |||
AK147171.1 | - | 4 | 466 | |||
AK160227.1 | - | 4 | 465 | |||
BC008233.1 | - | 4 | 452 | 2648048 | ||
BC064010.1 | - | 4 | 693 | 30244324 | ||
BC080837.1 | - | 4 | 447 | 5371708 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
