siRNA Details
Gene Symbol: Sgtb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144838.1 | NP_659087.1 | 7 | 763 | ||
GenBank mRNA | AK049820.1 | - | 7 | 801 | ||
AK082080.1 | - | 7 | 653 | |||
BC017611.1 | - | 7 | 763 | 4527683 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
