siRNA Details
Gene Symbol: Lrrc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146052.4 | NP_666164.1 | 2 | 842 | ||
GenBank mRNA | AK043406.1 | - | 2 | 729 | ||
BC019794.1 | - | 2 | 604 | 4481142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
