siRNA Details
Gene Symbol: Zfp503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145459.3 | NP_663434.2 | 3 | 2901 | ||
GenBank mRNA | AK032903.1 | - | 2 | 1450 | ||
AK046771.1 | - | 2 | 1918 | |||
AY281290.1 | - | 2 | 1971 | |||
AY281291.1 | - | 2 | 1945 | |||
BC058952.1 | - | 2 | 1818 | 5715786 | ||
BC066027.1 | - | 3 | 2901 | 6401144 |
