siRNA Details
Gene Symbol: Oxnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145460.2 | NP_663435.2 | 5, 6 | 759 | ||
GenBank mRNA | AK031978.1 | - | 4, 5 | 388 | ||
AK075924.1 | - | 4, 5 | 385 | |||
AK139884.1 | - | 5, 6 | 759 | |||
BC019806.1 | - | 5, 6 | 537 | 4018387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
