siRNA Details
Gene Symbol: Tmem55b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033271.4 | NP_001028443.1 | 7 | 839 | ||
GenBank mRNA | AK144251.1 | - | 7 | 839 | ||
AK152640.1 | - | 7 | 824 | |||
AK152658.1 | - | 7 | 824 | |||
AK159641.1 | - | 7 | 843 | |||
AK169026.1 | - | 3 | 2466 | |||
AK185482.1 | - | 3 | 206 | |||
AK205319.1 | - | 4 | 240 | |||
AK205696.1 | - | 4 | 238 | |||
AK213231.1 | - | 4 | 238 | |||
AK216843.1 | - | 3 | 206 | |||
AK219909.1 | - | 4 | 238 | |||
BC027355.1 | - | 7 | 824 | |||
BC039161.1 | - | 6 | 676 | |||
BC049945.1 | - | 6 | 696 | |||
BC058171.1 | - | 7 | 788 | |||
BC106163.1 | - | 7 | 818 | 4913757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
