siRNA Details
Gene Symbol: Khnyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027143.2 | NP_081419.1 | 8 | 1969 | ||
GenBank mRNA | AK032037.1 | - | 8 | 1941 | ||
AK033704.1 | - | 8 | 1970 | |||
AK047054.1 | - | 8 | 2515 | |||
AK047955.1 | - | 8 | 1938 | |||
AK122247.1 | - | 8 | 1981 | |||
AK136301.1 | - | 8 | 1997 | |||
BC116260.1 | - | 8 | 1956 | 40093805 | ||
BC116261.1 | - | 8 | 1956 | 40093806 |
